Learn More
BRAND™ Cover Glasses for Microscope Slides
Sfoglia Tutto Brand Prodotti
Click to view available options
Dimensioni (L x P):
18 x 18 mm
24 x 24 mm
24 x 40 mm
24 x 50 mm
24 x 60 mm
Forma:
Rectangle
Square
Dimensione della confezione:
pezzi 1000
pezzi 2000
Descrizione
- Thickness between 0.13 and 0.17mm
- Refractive index ne = 1.52 ± 0.01, Abbe number ve = 56.5 ± 0.5
- Fully automated production guarantees ready-for-use quality: clean, dust and grease-free, distortion-free
- Flatness within ± 3μm
- Pack quantity for square shape is 2000 (10 boxes of 200); pack quantity for rectangular shape is 1000 (10 boxes of 100)
Specifica
Specifica
| Forma | Square |
| Materiale | Borosilicate Glass |
| Spessore (sistema metrico) | #1 (0.13 to 0.16 mm) |
| Dimensioni (L x P) | 24 x 24 mm |
| Quantità | 2000 (10 Boxes of 200) |