Learn More
Cole-Parmer™ CPC™ Quick Disconnect Stainless Steel Sanitary Couplings
Integrate disposables with your existing stainless steel processing equipment.
Marca: Cole-Parmer™ 31604-30
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Attach these couplings directly to your sanitary hardware assemblies
USP Class IV materials. - Fully sterilizable.
- Both couplings and inserts are available with standard sanitary terminations to attach to your system with a sanitary gasket and tri-clover clamps.
- Choose from stainless steel or polysulfone inserts to mate to polysulfone bodies.
- All models feature USP Class VI platinum-cured silicone gaskets.
- Mix and match these bodies and inserts with the polysulfone quick-disconnect coupling elements for greater flexibility in your process system.
Specifica
Quick-Disconnect Insert, Stainless Steel, Sanitary Connector | |
Autoclavable | |
3/4 in. |
Sanitary x Quick-Disconnect | |
Stainless Steel | |
Autoclave or Gamma |