Learn More
Abeba™ Crawler ALU 34501 Shoes
ESD dissipative shoe is Sanitized™ treated. In cludes TPU sole with extremly high slip resistance.
Marca: Abeba™ 34501/43
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- CE EN ISO 20345:2011 S2 SRC
- Microfiber black/blue
- Aluminum toe cap
- Foamed PU midsole
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3577)
- Sanitized treated
- ESD dissipative according to DIN EN 61340
- Breathable lining with Silver ions
- Instep area with lacing
- Reflective material
- Upper one piece
Specifica
CE EN ISO 20345:2011 S2 SRC | |
Microfiber | |
Slip-resistant | |
34501 |
Black | |
43 | |
Polyurethane | |
Aluminum |