Learn More
Greiner Bio-One Provetta criogenica in polipropilene
Click to view available options
Capacità (sistema metrico):
2 mL
4 mL
Colore chiusura:
Blue
Green
Natural
Red
Verde
Yellow
Dimensione della confezione:
pezzi 300
pezzi 500
Descrizione
- Free of detectable human DNA, endotoxins, DNase and RNase
- Sterile, non-pyrogenic, non-cytotoxic, no heavy metals contained
- CE-marked (CE/IVD)
- As a saftety precaution Cryo.s should be stored in a freezer or exclusively in the gas phase over the liquid nitrogen
- Suitable for shipment by air
- With white, scratch-resistant writing area
- 150 inserts per case (for additional labelling on the cap)
- Suitable for storage at -20 °C, -80 °C and in the vapour phase of LN2
- Multiple cap colours and working volumes
- Ultra-pure medical grade, USP class VI certified polypropylene w/o leachables
- Accessory storages boxes and working racks available
Specifica
Specifica
| Product Type | Cryogenic Storage Tube |
| Colore chiusura | Red |
| Autoclavabile | Not Autoclavable |
| Capacità (sistema metrico) | 4 mL |
| Forma del fondo | Round (Starfoot) |
| Materiale | Polypropylene |
| Include | White writing area; cap inserts for labeling |
| Temperature Range (Metric) | -196°C to +100°C |
| Sterilità | Sterile |
| Graduated | Yes |
| Vedi altri risultati |