Learn More
Crystal Violet, ACS, 90+%, Thermo Scientific Chemicals
43.10€ - 113.00€
Identificatori chimici
CAS | 548-62-9 |
---|---|
Formula molecolare | C25H30ClN3 |
Molecular Weight (g/mol) | 407.986 |
Numero MDL | MFCD00011750 |
InChI Key | ZXJXZNDDNMQXFV-UHFFFAOYSA-M |
Sinonimo | Basic Violet 3; C.I. 42555 |
PubChem CID | 11057 |
ChEBI | CHEBI:41688 |
IUPAC Name | [4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride |
SMILES | CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
11308107
|
Thermo Scientific Alfa Aesar
022866.14 |
25 g |
43.10€
25g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11318107
|
Thermo Scientific Alfa Aesar
022866.22 |
100 g |
113.00€
100g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
Crystal Violet is used as an acid-base indicator, an alcohol denaturant, a biological stain, a textile dye and an indicator for copper salts. It is a component of gram staining in microorganisms and cell lines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identificatori chimici
548-62-9 | |
407.986 | |
ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
11057 | |
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride |
C25H30ClN3 | |
MFCD00011750 | |
Basic Violet 3; C.I. 42555 | |
CHEBI:41688 | |
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] |
Specifica
Crystal Violet | |
548-62-9 | |
MFCD00011750 | |
4077708 | |
Basic Violet 3; C.I. 42555 | |
ZXJXZNDDNMQXFV-UHFFFAOYSA-M | |
[4-[bis[4-(dimethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-dimethylazanium;chloride | |
11057 | |
407.99 | |
ACS |
∼215°C (decomposition) | |
C25H30ClN3 | |
UN3077 | |
14,4395 | |
Soluble in water and chloroform. Slightly soluble in alcohol and glycerol. | |
CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-] | |
407.986 | |
CHEBI:41688 | |
≥90% | |
Powder |
Sicurezza e movimentazione
GHS H Statement
H318-H351-H302
Causes serious eye damage.
Suspected of causing cancer.
Harmful if swallowed.
P201-P202-P264b-P270-P281-P301+P312-P305+P351+P338-P308+P313-P310-P330-P501c
H302-H318-H351
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 208-953-6
missing translation for 'rtecsNumber' : BO9000000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only