Learn More
BD Biosciences™ CS&T IVD Beads (CE/IVD)
Click to view available options
N. di test:
150 Tests
50 Tests
Dimensione della confezione:
Pezzo
Descrizione
- Intended for in vitro diagnostic use on BD FACSCanto™ II flow cytometers running BD FACSDiva™ software.
- Used to set up the cytometer, to perform daily performance quality control (QC), and to determine lyse/wash (LW) application settings.
- Dyed with fluorochromes that are excited by the cytometer's lasers.
Specifica
Specifica
| Content And Storage | Store vials at 2°C–8°C and protect from light. |
| N. di test | 50 Tests |
| Stabilità | The beads are stable until the date shown on the vial label when stored as directed. Do not use after the expiration date. After dilution, the beads are stable for 8 hours at 2°C–25°CC |
| Product Type | Fluorescent Polystyrene Beads |