Learn More
Sartorius Cubis™ II High-Capacity Micro Balance, 66P
The Cubis™ II range of high-capacity micro balances with a maximum load between 32 g and 111 g and a readability between 0.001 mg and 0.002 mg provide the ideal choice for a broad range of applications.
Marca:  Sartorius MCA66P-3CEU-D HWL
Dettagli aggiuntivi : Peso : 14.30000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17907380
  • 35539.00€ / Pezzo
Spedizione stimata: 11-09-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Ultra-high resolution with up to 61 million weighing steps (digits).
  • End-to-end data integrity.
  • 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management.
  • Cleanability, part of compliance.
  • Error-free operation.
  • Individual QApp workflows, motorized auto-leveling.
  • Allow upgrades for hardware features like automated draft shields or built-in ionizer.
  • Optional inner draft shield for best weighing performance.
Specifica
Specifica
MCA
Color Touchscreen
66P Weighing Module
3.5, 2.5 sec.
9.4 in.
11.9 in.
12, 61 g
0.001, 0.01 mg
MCA66P-3CEU-D HWL
Balance
240 mm
302 mm