Learn More
Thermo Scientific™ Culti-Loops™ Candida geochares ATCC™ 36852™
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Marca:  Thermo Scientific™ R4601401
Codice prodotto. 12938888
  • 223.00€ / pezzi 5
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifica
Specifica
Candida species
Candida geochares
Culti-Loops Quality Control Organism
Qualitative QC
CBS 6870 [CCRC 22394, IFO 10278, NRRL Y-17073]
36852TM*
Culti-Loops
Fungi
5 Loops/Pack
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Offerte speciali
Offerte speciali
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.