Learn More
Thermo Scientific™ Culti-Loops™ Optochin Test QC Set
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Marca: Thermo Scientific™ R4651108
Dettagli aggiuntivi : Peso : 0.01000kg Transport : UN number : 3373 Chem class : 6.2
Descrizione
Thermo Scientific™ Culti-Loops™ Optochin Test Set contains quality control organisms supplied in Culti-Loops format.
Culti-Loops are ready to use, disposable inoculating loops containing stabilized, preserved, viable microorganisms. The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media. Culti-Loops are recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Includes:
Streptococcus pneumoniae ATCC™ 6305™†
Streptococcus sanguis Type 1 ATCC™ 10556™†
Ordering Alerts:
Individually sealed in a foil pouch and packaged five loops per package.
Specifica
6305TM* | |
Culti-Loops | |
Organism Sets | |
5 Loops/Pack |
CLIA '88 Compliance Optochin Test QC Set/2 | |
Culti-Loops Quality Control Organism | |
Qualitative QC |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.