missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Tryptophan methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantità:
1 g
5 g
Descrizione
D-Tryptophan Methyl Ester Hydrochloride is an intermediate in the synthesis of Tryptophan derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Tryptophan Methyl Ester Hydrochloride is an intermediate in the synthesis of Tryptophan derivatives.
Solubility
Soluble in water (150 mg/ml), dimethyl sulfoxide and methanol (50 mg/ml).
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
D-Tryptophan Methyl Ester Hydrochloride is an intermediate in the synthesis of Tryptophan derivatives.
Solubility
Soluble in water (150 mg/ml), dimethyl sulfoxide and methanol (50 mg/ml).
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Specifica
Specifica
Materiale o nome chimico | D-Tryptophan methyl ester hydrochloride |
Punto di fusione | 213°C to 216°C |
Informazioni di solubilità | Soluble in water (150mg/ml),dimethyl sulfoxide and methanol (50mg/ml). |
Peso formulazione | 254.71 |
Percent Purity | 98% |
Quantità | 1 g |
Optical Rotation | −18° (c=5 in Methanol) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'