Learn More
Thermo Scientific™ Deionizer Cartridges
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Da utilizzare con (apparecchiatura):
B-Pure™
Bantam™ Deionizer, B-Pure™
Milli-Q Millipore Water Systems
Nanopure
Thermo Scientific™ Barnstead™ Bantam™ B-Pure™ or Nanopure™ Water Purification Systems as Indicated
Tipo di resina:
Activated Carbon
Extruded Carbon
Letto misto
Macroreticular, Activated Carbon
Mixed Bed
Mixed Bed, Activated Carbon
Mixed Bed, Ultrapure, Oxygen and Organic Removal
Oxygen Removal Mixed Bed
Oxygen Removal Resin
Porous Strong Anion Exchange
Porous Strong Ion Exchange
Strong Anion Exchange
Strong Cation Exchange
Super-C Carbon
Two Bed Ion Exchange
Ultrapure, Oxygen Removal
Ultrapure, Oxygen and Organic Removal
Da utilizzare con (applicazione):
Anion Removal
Cation Removal
High capacity, two-bed ion exchange
Ion Exchange, Ultrapure
Organic Removal
Oxygen Removal
Oxygen and demineralization
Pretreatment
Pretreatment Macropure
Ultrapure
Ultrapure and Organic Removal
Ultrapure and Oxygen Removal Resin
Ultrapure, Oxygen and Organic Removal
Dimensione della confezione:
1 unità
Pezzo
Capacità:
1680 grains
1760 grains
18 g
2000 gal.
275 grains/2000 gal.
275 grains/9 g
30 g
3170 grains
365 grains/2000 gal./12 g
455 grains/30 g
5000 gal.
5000 gal. (as CaCO3)
550 grains
785 grains/1000 gal.
875 grani
Descrizione
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
- Choice of full or half cartridges
Specifica
Specifica
| Descrizione | High Temp, Ultrapure, Oxygen and Organic Removal Cartridge |
| Tipo di resina | Ultrapure, Oxygen Removal |
| Da utilizzare con (applicazione) | Ultrapure, Oxygen and Organic Removal |
| Da utilizzare con (apparecchiatura) | B-Pure™ |
| Tipo | Deionizer Cartridge |