missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipotassium Hydrogen Orthophosphate Trihydrate, Extra Pure, SLR, Fisher Chemical™
65.00€ - 99.30€
Identificatori chimici
CAS | 16788-57-1 |
---|---|
Formula molecolare | H7K2O7P |
Molecular Weight (g/mol) | 228.22 |
Numero MDL | MFCD00149926 |
InChI Key | XQGPKZUNMMFTAL-UHFFFAOYSA-L |
Sinonimo | dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate |
PubChem CID | 16217523 |
IUPAC Name | dipotassium;hydrogen phosphate;trihydrate |
SMILES | O.O.O.[K+].[K+].OP([O-])([O-])=O |
Descrizione
Identificatori chimici
16788-57-1 | |
228.22 | |
XQGPKZUNMMFTAL-UHFFFAOYSA-L | |
16217523 | |
O.O.O.[K+].[K+].OP([O-])([O-])=O |
H7K2O7P | |
MFCD00149926 | |
dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate | |
dipotassium;hydrogen phosphate;trihydrate |
Specifica
16788-57-1 | |
8.5 to 9.6 | |
H7K2O7P | |
dipotassium hydrogen phosphate trihydrate, unii-rjk174x3tz, potassium hydrogenphosphate trihydrate, potassium phosphate dibasic trihydrate, potassium hydrogen phosphate trihydrate, potassium phosphate, dibasic, trihydrate, phosphoric acid, dipotassium salt, trihydrate, rjk174x3tz, dipotassium phosphate trihydrate | |
O.O.O.[K+].[K+].OP([O-])([O-])=O | |
228.22 | |
228.23g/mol |
White | |
Solid | |
MFCD00149926 | |
XQGPKZUNMMFTAL-UHFFFAOYSA-L | |
dipotassium;hydrogen phosphate;trihydrate | |
16217523 | |
Plastic powder jar |