Learn More
Esculape Laboratoire™ Spray Diphoterine
Use Esculape Laboratoire™ Diphoterine Spray Neutralizing Agent. This hypertonic solution attracts the chemical mixture product that has penetrated the tissues and neutralizes it;by osmotic effect.
Marca: Esculape Laboratoire™ 8205210
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Sterile polyvalent
- Hypertonic solution
- Used to wash face and body
- 100mL
Specifica
H 170 x diam. 45 mm | |
170 mm |