Learn More
KGW-Isotherm Dish-Shaped Dewer Flasks, Aluminum Cover
Reliable and economical Dewar flasks according to DIN EN ISO 16496. KGW-Isotherm Dish-Shaped Dewer Flasks, Aluminum Cover pairs a bowl shaped glass flask in a protective casing made of stucco aluminum for easy handling. Lid sold separately.
Marca: KGW-Isotherm 1192
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Made of borosilicate glass according to ISO 3585
- Structured aluminum cover: antimagnetic and stainless
- Fully silvered
- Applications:
- tempering of round bottom flasks
- cooling of small samples
- suitable as a hot or cold bath
- suitable on magnetic stirrer
- Chemical characteristics:
- Hydrolytic resistance: according to ISO 719 (98°C)
- Hydrolytic resistance: according to ISO 720 (121°C)
- Acid resistance: according to ISO 1776
- Alkaline resistance: according to ISO 695-A2
- Temperature range:
- Glass refill from -200°C to + 150°C, for a short time 200°C
- Casing with mounting rubber and Latizell buffer from -10°C to + 60°C
Specifica
Silver | |
95-110 mm | |
50 mm | |
SCH 9 CAL | |
120 mL | |
Fully silvered |
77 mm | |
Tempering, Cooling, Warming, Stirring | |
90 mm | |
Aluminum | |
Borosilicate Glass |