Learn More
Epredia™ Disposable Base Molds
Sfoglia Tutto Epredia Prodotti
Click to view available options
Dimensioni (L x P):
7 x 7 mm
15 x 15 mm
24 x 24 mm
30 x 24 mm
37 x 24 mm
Dimensione della confezione:
pezzi 500
Descrizione
- Cleaning associated with metal molds is not required
- Transparency facilitates tissue orientation
Ordering Information:
- Sold in cases of four boxes.
Specifica
Specifica
| Product Type | Base Mold |
| Materiale | Plastic |
| Dimensioni (L x P) | 24 x 24 mm |
| Monouso | Single-use |