Learn More
3M™ Semimaschera
Click to view available options
Tipo cartuccia:
FFABEK1P3 R D
Dimensione della confezione:
Pezzo
Descrizione
Simple, lightweight, well-balanced respirator offers reliable respiratory protection against a range of gases, vapors, and particulates.
- No parts assembly or maintenance required
- Unique bonded carbon technology allows the respirator to sit close to the face
- Low profile design allows the respirator to fit under many other types of protective equipment, providing an undisturbed field of vision and a more comfortable worklife
- Exhalation valve centrally positioned for effective removal of heat and moisture
- Can be used until damaged, particulate filters clog, gas filters become saturated or after one month
- Fine dusts
- Oil and water based mists
- Organic gases or vapours with good warning properties and boiling point above 65°C
- EN 405:2002
- Maximum usage limitations:
- Gases/vapours: APF - 10 x WEL
- Particulates: APF - 20 x WEL
- CE marked
Specifica
Specifica
| Tipo cartuccia | FFABEK1P3 R D |
| Certificazione/Conformità | Marchio CE, EN 405: 2002, Classe FFABEK1P3D |
| Product Type | Respiratore con semimaschera facciale |
| Da utilizzare con (applicazione) | Protezione delle vie respiratorie da polveri sottili, nebbie a base acquosa e oleosa, gas e vapori organici con buone garanzie di allerta e punto di ebollizione oltre i 65 °C, gas inorganici, gas acidi e ammoniaca |
| Modello | Serie 4000 |
| Quantità | 1 ciascuno |