Learn More
Swann-Morton™ Disposable Scalpels, Non-Sterile, Bulk Pack with Guard, No. 15
Click to view available options
Quantità:
100/Box
Dimensione della confezione:
pezzi 10
Descrizione
- Small, curved cutting edge.
- Most popular blade shape.
- Ideal for making short and precise incisions.
- Used in a variety of surgical procedures including the excision of a skin lesion or recurrent sebaceous cyst and for opening coronary arteries.
- Fits handles 3, 3L, 3 graduated, 5B, 7, 9, B3, B3L.
Specifica
Specifica
| Materiale della lama | Stainless Steel |
| Monouso | Single-use |
| Sterilità | Non-sterile |
| Quantità | 100/Box |
| Tipo di imballaggio | Bulk Packaged |