missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals DL-Aspartic acid, 98+%
Click to view available options
Quantità:
100 g
500 g
2500 g
Descrizione
DL-Aspartic acid is used in studies on factors and conditions that enhance semen and sperm quality. Its conjugate base D-aspartate has potential use as a therapeutic agent in the treatment of schizophrenia-related symptoms.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Aspartic acid is used in studies on factors and conditions that enhance semen and sperm quality. Its conjugate base D-aspartate has potential use as a therapeutic agent in the treatment of schizophrenia-related symptoms.
Solubility
Soluble in water, and hydrochloric acid. Insoluble in ether, and alcohol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
DL-Aspartic acid is used in studies on factors and conditions that enhance semen and sperm quality. Its conjugate base D-aspartate has potential use as a therapeutic agent in the treatment of schizophrenia-related symptoms.
Solubility
Soluble in water, and hydrochloric acid. Insoluble in ether, and alcohol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
Specifica
Materiale o nome chimico | DL-Aspartic acid |
Punto di fusione | >300°C (decomposition) |
Beilstein | 774618 |
Informazioni di solubilità | Soluble in water,and hydrochloric acid. Insoluble in ether,and alcohol. |
Peso formulazione | 133.1 |
Percent Purity | ≥98% |
Quantità | 500 g |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'