Learn More
Parker Domnick Hunter™ Nitrogen Gas Generator
Click to view available options
Tipo:
Nitrogen Gas Generator
Dimensione della confezione:
Pezzo
Descrizione
- Complete 'Plug and Play' system specifically designed for critical analytical applications
- Produces a continuous supply of 99.999% purity nitrogen 24 hours a day
- Integral oil free compressor, with noise reduction technology
- Eliminate inconvenient and potentially dangerous nitrogen cylinders
- Compact, reliable with minimal operator attention and maintenance
- Phthalate-free componentry
Specifica
Specifica
| Portata | 0.75 L/min. |
| Dimensioni uscita | 1/8in. |
| Tensione | 240 V |
| Purezza dell'azoto | >99.999% |
| Warranty | 2Years |
| Frequenza | 50 Hz |
| Tipo | Nitrogen Gas Generator |