Learn More
Socorex™ Dosys™ Basic Pistol Grip Automatic Syringe
Dose with excellent reproducibility using a pistol grip handle automatic syringe. Socorex™ Dosys™ Basic Pistol Grip Automatic Syringe fits either hand comfortably and requires limited effort for efficient work even during long dispensing sessions.
Marca: Socorex™ 172.0502
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Pistol grip handle.
- Automatic syringe.
- Without valve system.
- Luer lock nozzle.
- Fits either hand.
- Requires limited effort.
- High reproducibility due to safety locking nut on a micrometric screw.
- High-precision stainless steel plunger.
- Ground glass barrel.
- Smooth plunger travel and tightness.
- Glass barrel protected by PVC sleeve (not autoclavable).
- Easy disassembly for efficient cleaning and parts replacement if necessary.
Specifica
Autoclavable | |
Syringe, PVC protection sleeve, operating instructions. | |
0.1 mL | |
1/ea. | |
0.3 to 2 mL |
Borosilicate Glass (Barrel), Stainless Steel (Plunger) | |
Automatic Syringe, Pistol Grip | |
Dosys™ Basic 172 | |
<0.6% to <0.4% | |
Yes |