Learn More
Socorex™ Dosys™ Classic Pistol Grip Automatic Syringe
Codice prodotto. 10274462
Click to view available options
Volume (sistema metrico):
0.025 to 0.3 mL
0.5 to 5 mL
1 to 10 mL
5 to 20 mL
Dimensione della confezione:
Pezzo
Codice prodotto. 10274462
Marca: Socorex™ 173.0520
Dose with excellent reproducibility using a pistol grip handle automatic syringe. Socorex™ Dosys™ Classic Pistol Grip Automatic Syringe fits either hand comfortably and requires limited effort for efficient work even during long dispensing sessions.
  • Pistol grip handle.
  • Self-refilling automatic syringe.
  • Integrated, Luer lock valve system.
  • Fits either hand.
  • Requires limited effort.
  • High reproducibility due to safety locking nut on a micrometric screw.
  • High-precision stainless steel plunger.
  • Ground glass barrel.
  • Smooth plunger travel and tightness.
  • Glass barrel protected by PVC sleeve (not autoclavable).
  • Easy disassembly for efficient cleaning and parts replacement if necessary.
Specifica
Autoclavabile Autoclavable
Materiale Borosilicate Glass (Barrel), Stainless Steel (Plunger)
Include Syringe, 1M silicone feed tube, sinker, PVC protection sleeve, aspiration and vent cannulas, set of spares for valve, operating instructions.
Product Type Automatic Syringe, Pistol Grip
Graduazioni 1.0 mL
Modello Dosys™ Classic 173
Quantità 1/ea.
Precisione <0.5% to <0.4%
Volume (sistema metrico) 5 to 20 mL
Tipo di collegamento Luer Lock
Graduated Yes