missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC Columns

Codice prodotto. 30040297 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Lunghezza (sistema metrico):
150 mm
500 mm
750 mm
Product Line:
Double nanoViper
Single nanoViper
Dimensione della confezione:
1 unità
Pezzo
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 30040297

Marca: Thermo Scientific™ DNV150150PN

Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Separate challenging peptide mapping samples with Thermo Scientific™ Double nanoViper™ PepMap™ Neo UHPLC columns.

Separate challenging peptide mapping samples with Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC columns. These columns feature flexible connectivity, high reproducibilty, and excellent separation performance. Our PepMap Neo columns are packed to higher pressure and provide 1500 bar pressure capability, improved column-to-column consistency, and increased efficiency. The column media is manufactured and selected to exacting standards and packed at high pressure, resulting in enhanced peak symmetry, resolution, and column-to-column reproducibility that allows you to reliably obtain greater sample coverage.

Benefits of PepMap Neo Columns

  • Brilliant Connectivity — Double nanoViper™ columns connect with ease.
  • Brilliant Reproducibility — Precise manufacturing protocols for packing media improve column-to-column consistency for reproducible analysis every day.
  • Brilliant Separation — Ultra-high pressure packing increases column efficiency, allowing you to run at up to 1500 bar for enhanced peak symmetry and resolution, providing higher sample capacity for more sample insights.
TRUSTED_SUSTAINABILITY

Specifica

Formato colonna Analytical Column, Double nanoViper
Diametro (sistema metrico) 150 µm
Lunghezza (sistema metrico) 150 mm
Pressione max 22,000 psi (1500 bar)
Dimensione particelle 2 μm
pH 2 to 8
Dimensione del poro 100 Å
Surface Area 450 m2/g
Temperatura 60 °C
Tipo USP L1
Fase stazionaria C18
Product Line Double nanoViper
Materiale di impaccamento diossido di silicio ultrapuro completamente poroso, sferico
Carico di carbonio 15%
Tipo di colonna A fase inversa
Con tappo
Fase A fase inversa
Descrizione Double nanoViper PepMap Neo 2 µm C18 150 µm x 150 mm
Da utilizzare con (applicazione) Peptide Mapping, LC-MS
Vedi altri risultati Mostra meno risultati
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.