Learn More
Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC Columns
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
Separate challenging peptide mapping samples with Thermo Scientific™ Single nanoViper™ and Double nanoViper™ PepMap™ Neo UHPLC columns. These columns feature flexible connectivity, high reproducibilty, and excellent separation performance. Our PepMap Neo columns are packed to higher pressure and provide 1500 bar pressure capability, improved column-to-column consistency, and increased efficiency. The column media is manufactured and selected to exacting standards and packed at high pressure, resulting in enhanced peak symmetry, resolution, and column-to-column reproducibility that allows you to reliably obtain greater sample coverage.
Benefits of PepMap Neo Columns
- Brilliant Connectivity — Double nanoViper™ columns connect with ease.
- Brilliant Reproducibility — Precise manufacturing protocols for packing media improve column-to-column consistency for reproducible analysis every day.
- Brilliant Separation — Ultra-high pressure packing increases column efficiency, allowing you to run at up to 1500 bar for enhanced peak symmetry and resolution, providing higher sample capacity for more sample insights.
Specifica
Specifica
| Formato colonna | Analytical Column, Single nanoViper |
| Diametro (sistema metrico) | 75 µm |
| Lunghezza (sistema metrico) | 750 mm |
| Pressione max | 22,000 psi (1500 bar) |
| Dimensione particelle | 2 μm |
| pH | 2 to 8 |
| Dimensione del poro | 100 Å |
| Surface Area | 450 m2/g |
| Temperatura | 60 °C |
| Tipo USP | L1 |
| Vedi altri risultati |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.