Learn More
Techspray Double-Tip Pointed Cotton Swab
Tightly wrapped, pre-cleaned cotton ensures the least amount of extractable contaminants. Pointed tip and rigid head provide precision, but aggressive cleaning.
Marca: Techspray 2313-30PK
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
- Tightly wrapped cotton
- Precision pointed tip
- Low extractable
- Solvent resistant
Specifica
Pointed | |
Cotton | |
3.2 mm | |
30 |
Composite | |
Double-Tip Swab | |
12 mm |