Learn More
Thermo Scientific™ DRI™ Creatinine-Detect™ Specimen Validity Test Calibrators
Rise to the challenge of combating the use of additives and adulterants to mask controlled substances with Thermo Scientific™ DRI™ Creatinine-Detect™ Specimen Validity Test Calibrators.
Marca: Thermo Scientific™ 100272
Description
Combat the use of additives and adulterants to mask controlled substances in urine samples with Thermo Scientific™ DRI™ Creatinine-Detect™ Specimen Validity Test Calibrators. Liquid and ready-to-use, these calibrators can be used on a variety of general chemistry analyzers.
The use of additives and adulterants to mask controlled substances in urine samples is a difficult and ongoing challenge to drugs of abuse screening. We offer the industry's most extensive menu of detection tests. As new adulterant trends sweep through the testing pool, rely on our fast-track product development team to deliver a timely, economical detection solution. The adulteration assays provide a complete and automated way to test for sample integrity and adulteration.
- Complete and automated way to test for sample integrity and adulteration
- Associated controls offered in convenient sizes
Spécification
2°C to 8°C | |
2.0, 20 mg/dL | |
Urine | |
2 x 25 mL |
Creatinine Assay | |
Creatinine | |
Calibrator |