Learn More
Thermo Scientific™ Drums for Digital Cel-Gro Tissue Culture Rotators
Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V
Marca:  Thermo Scientific™ 88882152
Dettagli aggiuntivi : Peso : 1.05000kg
Codice prodotto. 17308282
  • 174.00€ / Pezzo
Spedizione stimata: 05-11-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V
Specifica
Specifica
Rotating Drum
290 mm
10/15 mL, 5/7 mL tubes
30
Stainless Steel
10 x 10/15 mL and 20 x 5/7 mL rotator drum
Accessory for Digital Cel-Gro Tissue Culture Rotator, 120V (88882015) and Digital Cel-Gro Tissue Culture Rotator 230V (88882016)