Learn More
Lenz Laborglasinstrumente™ Palloni a tre colli in vetro borosilicato DURAN™ con colli laterali inclinati
Especially made for distillation, reactions and to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Angled are manufactured from durable DURAN™ borosilicate glass.
Marca:  Lenz Laborglasinstrumente™ 03314482
Dettagli aggiuntivi : Peso : 0.01000kg
Codice prodotto. 10080211
  • 408.10€ / Pezzo
Spedizione stimata: 22-10-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Acc. to DIN 12394
  • Two side necks at 20° angles
  • With conical ground joints, featuring high taper accuracy and excellent tightness
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifica
Specifica
5 L
Borosilicate Glass, Duran Glass
29/32 NS Side Neck, 29/32 NS Center Neck
Round