Learn More
Lenz Laborglasinstrumente™ Tubi filettati in vetro Duran™
Provides for quick and easy leak-tight connections to apparatus in the laboratory. Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone are manufactured from durable DURAN™ borosilicate glass.
Marca: Lenz Laborglasinstrumente™ 01311814
Dettagli aggiuntivi : Peso : 0.00900kg
Descrizione
- Acc. to DIN 12257
- Corrosion-free and resistant to thermal shock
- With conical joint, featuring high taper accuracy and excellent tightness
Specifica
Provette con filettatura | |
DIN 12257 | |
Vetro Duran | |
Confezione da 1 |