Learn More
DWK Life Sciences DURAN™ KPG™ Stirrer Bearing, interchangeable, with standard ground cone, HP, HQ
KPG bearing sleeve is a precisely fitting holder for guiding the KPG agitator shaft.
Marca:  DWK Life Sciences 245235504
70.78 EUR valido fino al 2024-12-31
Usa il codice promo "23258" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.03400kg
Codice prodotto. 10008230
  • 74.50€ / Pezzo
Spedizione stimata: 11-10-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
KPG agitators consist of a KPG agitator shaft and a KPG bearing sleeve. KPG bearing parts are manufactured with high accuracy and uniformity. The specified tolerances for nominal diameter and fit are maintained by careful checks. This guarantees the interchangeability of the KPG bearing sleeves and KPG stirrer shafts. Due to the low bearing clearance between the running surfaces, almost absolute sealing and completely quiet running can be achieved with suitable lubrication. KPG agitators are an alternative to magnetic stirrers and are used for reaction vessels that are not accessible from below when very viscous reaction mixtures are to be stirred or a high stirring speed is required.
  • High accuracy and uniformity within defined tolerances for nominal diameter and fit
  • Tight closure due to standard ground joint
  • Ground and polished bearing surface
  • Interchangeable
Specifica
Specifica
1 Ea.
Stirring of liquids (for syntheses); Stirring at high stirring speed; Mounting support
10 mm
HP 10, NS 24/29