Learn More
DWK Life Sciences DURAN™ Stopcock, with PTFE Spindle, for threaded outlets, type MOBILEX (GL 32)
The PTFE spindle valve is used for airtight sealing of a desiccator.
Marca: DWK Life Sciences 247970305
36.86 EUR valido fino al 2024-12-31
Usa il codice promo "23258" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.04600kg
Descrizione
The PTFE spindle valve is used for airtight sealing of a desiccator. For this purpose, the spindle tap is connected to the standard ground joint of the desiccator cover. The very small, punctual opening in the lower part of the spindle valve allows a slow inflow of gases or air when the valve is opened, thus reducing a strong turbulence inside the desiccator.
- Small grinding opening of the spindle valve allows a slow inflow of gases or air and thus prevents excessive turbulence
- Tight closure due to standard ground joint
- For threaded outlets of MOBILEX type desiccator (GL 32)
Specifica
8 mm | |
Borosilicate Glass 3.3, PTFE | |
Shutting off or diverting and dosing gases and vapors; Closure of desiccators |