Learn More
Abeba™ Dynamic 7313 Shoes
Special respiration absorbing lining allows the foot to breathe. Includes TPU sole with high slip resistance.
Marca: Abeba™ 7313/42
44.27 EUR valido fino al 2024-07-31
Usa il codice promo "21674" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.94000kg
Descrizione
- CE EN ISO 20347:2012 OB A, E, FO SRC
- Smooth leather white/red
- Slip resistance TPU outsole sole
- Replaceable acc Wave insole (article no. 3574)
- Sanitized treated
- Antistatic
- Heel strap foldable, adjustable
Specifica
CE EN ISO 20347:2012 OB A, E, FO SRC | |
Leather | |
Slip-resistant | |
7313 |
White | |
42 | |
Polyurethane |