Learn More
IBA Lifesciences pASG-IBA E. coli Expression Vector with Secretion
Sfoglia Tutto IBA Lifesciences Prodotti
Descrizione
-
The pASG-IBA vectors are designed for expression of target proteins without affinity tag in E. coli.
-
The vector carries an ampicillin resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, and the inducible tetracycline promoter/operator for the regulated expression of proteins.
-
The vectors can be combined with any E. coli strain since the tet-promoter works independently of the genetic background of E. coli.
-
Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity.
Please note that cloning into pASG-IBA Acceptor Vectors compulsorily requires the restriction enzyme Esp3I since no other MCS for the integration of a gene of interest is available. In addition to the direct cloning of the gene of interest into pASG-IBA vectors with Esp3I, another option via a so-called Entry-Vector is possible.
Specifica
Specifica
| Agente inducente | Inducible with anhydrotetracycline |
| Gene marcatore | Ampicillin |
| Promotore | Tet |
| Product Type | Vector |
| Content And Storage | Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage |
| Protein Tag | Strep-tag II |
| Quantità | 5 μg |
| Vettore | pASG-IBA4 |
| Da utilizzare con (applicazione) | ompA for periplasmic secretion |