missing translation for 'onlineSavingsMsg'
Learn More

EC 23, Tocris Bioscience™

Codice prodotto. 18752694
Click to view available options
Quantità:
10 mg
50 mg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Synthetic retinoid; induces differentiation of stem cells

Photostable synthetic retinoid; analog of ATRA (Cat. No. 0695). Displays similar activity to ATRA when tested on the embryonal carcinoma stem cell model TERA2.cl.SP12. Potently induces neural differentiation in human pluripotent embryonic stem cells.

Identificatori chimici

CAS 104561-41-3
Formula molecolare C23H24O2
Molecular Weight (g/mol) 332.443
InChI Key OQVLOWLEEHYBJH-UHFFFAOYSA-N
Sinonimo vsdtydjdjj, unii-vsdtydjdjj, 4-2-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl ethynyl benzoic acid, 4-2-1,1,4,4-tetramethyltetralin-6-yl ethynyl benzoic acid, 4-2-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl ethynyl, 4-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl ethynyl benzoic acid, 4-2-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl ethynyl benzoic acid, 4-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-ylethynyl benzoic acid, 4-5,6,7,8-tetrahydro-5,5,8,8-tetramethylnaphth-2-yl-ethynyl-benzoic acid, 4-5,6,7,8-tetrahydro-5,5,8,8-tetramethylnaphth-2-yl ethynyl-benzoic acid
PubChem CID 10314719
IUPAC Name 4-[2-(5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethynyl]benzoic acid
SMILES CC1(CCC(C2=C1C=CC(=C2)C#CC3=CC=C(C=C3)C(=O)O)(C)C)C

Specifica

Materiale o nome chimico EC 23
Quantità 10 mg
Peso formulazione 332.44
Percent Purity >99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'