missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD ELISPOT Sets
Click to view available options
Antigene:
Granzyme B
IFN-gamma
IL-10
IL-2
IL-4
IL-5
TNF
Cross-reattività:
Human
Mouse
Product Type:
Human Granzyme B ELISPOT set
Human IFN-gamma ELISPOT set
Human IL-10 ELISPOT set
Human IL-2 ELISPOT set
Human IL-4 ELISPOT set
Human IL-5 ELISPOT set
Human TNF ELISPOT set
Mouse IFN-gamma ELISPOT set
Mouse IL-2 ELISPOT set
Mouse IL-4 ELISPOT set
Mouse IL-5 ELISPOT set
Mouse TNF ELISPOT set
Dimensione della confezione:
Pezzo
Descrizione
Includes
Unlabeled capture antibody (no azide/low endotoxin format), Biotinylated detection antibody, Streptavidin-HRP, 10 ELISPOT plates, Certificate of Analysis providing lot-specific optimal reagent concentrations
- High sensitivity
- High throughput, high content analysis
- Minimal volume of biological material required
- Suitable for frozen/thawed biological samples
- Compatibility with other assays—cells analyzed by BD ELISPOT Sets can be used for cloning, proliferation assays, flow cytometry, or other methods of analysis
Specifica
Specifica
| Antigene | IL-2 |
| Cross-reattività | Mouse |
| Include | Unlabeled capture Antibody, detection Antibody, Streptavidin HRP, 10 ELISPOT plates |
| Quantità | 10 x 96 Tests |
| Content And Storage | 4°C |
| Da utilizzare con (applicazione) | Cloning, proliferation assays, flow cytometry or other methods of analysis |
| Product Type | Mouse IL-2 ELISPOT set |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto