Learn More
Sartorius Entris™ II Advanced Precision Balances
Sfoglia Tutto Sartorius Prodotti
Click to view available options
Capacità:
220 g
320 g
420 g
620 g
820 g
1.2 kg
2.2 kg
3.2 kg
4.2 kg
5.2 kg
6.2 kg
8.2 kg
10.2 kg
12.2 kg
Leggibilità:
1 mg
10 mg
100 mg
Legal for Trade:
Class I
Class II
Class III
Calibrazione:
Internal
Unit Size:
Pezzo
Description
- Entris™ II Advanced balances - Quality, Value, and Consistency
- Accurate weighing results that can be trusted time after time
- Simplifies balance leveling with built-in, real-time level support
- Increases reliability of weighing results by using integrated protection systems
- Controlled access to balance settings with user management
- Gapless documentation thanks to CalAuditTrail
Specifications
Specifications
| Certificazione/Conformità | CE Certificate, CSA for Canada |
| Tipo di collegamento | D-sub connector, 9-pin; USB UTL; USB Device |
| Descrizione | With Internal Calibration |
| Capacità | 320 g |
| Tipo di display | Intuitive Graphic Touch Technology |
| Leggibilità | 1 mg |
| Materiale | Glass, PBT, Stainless Steel |
| Modello | Electronic Precision Balance |
| Tempo di stabilizzazione | ≤1.0 seconds |
| Tipo | Balance |
| Show More |