Learn More
Sartorius Entris™ II Precision Balance, Internal Adjustment
Sfoglia Tutto Sartorius Prodotti
Click to view available options
Capacità:
220 g
320 g
420 g
620 g
650 g
820 g
1200 g
2200 g
3200 g
4200 g
5200 g
6200 g
8200 g
Leggibilità:
1 mg
10 mg
100 mg
1000 mg
Modello:
CEU Electronic Toploading Balance
Dimensione della confezione:
1 unità
Descrizione
- No matter what you're weighing, the new Entris™ II balance is always the right choice.
- Offering unrivalled value and backed by almost 150 years of German engineering expertise, the Entris™ II comes in two product lines, so you can find the balance that meets your specific weighing needs.
- With over 40 models from which to choose, you will surely find a model to meet your basic weighing needs.
Specifica
Specifica
| Certificazione/Conformità | CEU |
| Tipo di collegamento | RS232, USB C |
| Capacità | 3200 g |
| Tipo di display | Touchscreen |
| Leggibilità | 10 mg |
| Materiale | PBT, Glass, Stainless Steel |
| Modello | CEU Electronic Toploading Balance |
| Tempo di stabilizzazione | ≤1.0 sec. |
| Tipo | Precision Balance |
| Requisiti elettrici | 100 – 240 VAC (±10%), 50 - 60 Hz, 0.2 A |