missing translation for 'onlineSavingsMsg'
Learn More

(Ethylthio)acetic acid, 97%, Thermo Scientific Chemicals

Codice prodotto. 11351697
Click to view available options
Quantità:
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 627-04-3
Formula molecolare C4H8O2S
Molecular Weight (g/mol) 120.166
Numero MDL MFCD00021769
InChI Key VJIKFWJCVWFZIN-UHFFFAOYSA-N
Sinonimo ethylthio acetic acid, acetic acid, ethylthio, ethylthioacetic acid, ethylmercapto acetic acid, carboxymethyl ethyl sulfide, s-carboxymethyl ethyl mercaptan, 2-ethylthio acetic acid, unii-m3d5y724hz, acetic acid, 2-ethylthio, ethylmercaptoacetic acid
PubChem CID 69387
IUPAC Name 2-ethylsulfanylacetic acid
SMILES CCSCC(=O)O

Specifica

Materiale o nome chimico (Ethylthio)acetic acid
Punti di ebollizione 117°C to 118°C (11 mmHg)
Indice di rifrazione 1.486
Quantità 25 g
Numero UN UN3265
Beilstein 1743038
Peso formulazione 120.18
Percent Purity 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'