Learn More
Thermo Scientific™ Eutech™ DO 6+ Dissolved Oxygen Meter ECDO601PLUSK
Descrizione
Dissolved Oxygen (DO) meters and probes measure the amount of gaseous oxygen dissolved in a solution. In theory, the amount of DO in a solution is dependent on 3 factors, namely temperature, salinity and atmospheric pressure. The measurement of DO requires a special DO electrode that is made up of an anode, a cathode, electrolyte solution and a gas permeable membrane.
The Eutech™ DO 6+ Dissolved Oxygen Meter ECDO601PLUSK comes with 1m Cable Electrode ECDO6HANDY3M, 2 Assembled Membrane Cap Housing, 1 Refilling Electrolyte (10 ml) & Carrying Kit Set.
Features:
- User Customisation for user's flexibility like toggling of various modes from ppm, mg/l to % saturation, view electrode diagnosis, etc.
- Calibration can be performed at 100% and/or 0% and mg/l or ppm against a known solution
- Automatic Temperature Compensation (ATC) guarantees the highest accuracy under varying temperature condition
- Galvanic Cell allows instantaneous measurements, producing quick responses and eliminates polarisation; requires minimal maintenance
- Automatically Compensates Salinity and Barometric Pressure after manual input for increased accuracy
- Hold Function momentarily freezes reading for easy viewing
- Auto-Power Off saves battery power after non-use
- Self-Diagnostic with message codes for easy troubleshooting
- Large Custom LCD provides optimum viewing even at a distance
- Protective Rubber Boot shields meter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
- Rugged Carrying Case (Optional) provides handy on-site meter calibration and measurement
Specifica
Specifica
| Descrizione | Eutech DO 6+ Dissolved Oxygen Meter with 1m Cable Electrode ECDO6HANDY3M, 2 Assembled Membrane Cap Housing, 1 Refilling Electrolyte (10 ml) & Carrying Kit Set |
| Include | Self-diagnostic; Electrode characteristics; Hold & Auto-Power Off after20 minutes |
| Correzione della pressione barometrica | 500 to 1499 mmHg or 66.6 to 199.9 kPA |
| Modalità di calibrazione | Up to 2 Calibration points; 100% in air and/or 0% in known solution |
| Intervallo di temperatura (sistema metrico) | 0.0°C to 50.0°C |
| Risoluzione (temperatura) | 0.1°C (0.1°F) |
| Precisione (Temperatura) | ± 0.3 °C (0.5 °F) |
| Tipo di display | LCD |
| Intervallo (RDO) | 0 to 20.00 mg/l or ppm |
| Metodi | Automatic correction after manual input |
| Vedi altri risultati |