Learn More
Thermo Scientific™ Eutech™ DO 6+ Dissolved Oxygen Meter
Descrizione
Dissolved Oxygen (DO) meters and probes measure the amount of gaseous oxygen dissolved in a solution. In theory, the amount of DO in a solution is dependent on 3 factors, namely temperature, salinity and atmospheric pressure. The measurement of DO requires a special DO electrode that is made up of an anode, a cathode, electrolyte solution and a gas permeable membrane.The Eutech™ DO 6+ Dissolved Oxygen Meter comes with 3m Cable Electrode ECDO6HANDY3M, 2 Assembled Membrane Cap Housing, 1 Refilling Electrolyte (10 ml) & Carrying Kit Set.
Features:
- User Customisation for user's flexibility like toggling of variousmodes from ppm,mg/l to % saturation, view electrode diagnosis, etc.
- Calibration can be performed at 100% and/or 0% andmg/l or ppm against a known solution
- Automatic Temperature Compensation (ATC) guarantees the highest accuracy under varying temperature condition
- Galvanic Cell allows instantaneous measurements, producing quick responses and eliminates polarisation; require sminimal maintenance
- Automatically Compensates Salinity and Barometric Pressure aftermanual input for increased accuracy
- Hold Functionmomentarily freezes reading for easy viewing
- Auto-Power Off saves battery power after non-use
- Self-Diagnostic withmessage codes for easy troubleshooting
- Large Custom LCD provides optimum viewing even at a distance
- Protective Rubber Boot shieldsmeter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
- Rugged Carrying Case (Optional) provides handy on-sitemeter calibration andmeasurement
Specifica
Specifica
| Include | 3 m Cable Electrode, 2 Assembled Membrane Cap Housing, 1 Refilling Electrolyte (10 ml), Carrying Kit Set |
| Tipo | Dissolved Oxygen Meter |