Learn More
Invitrogen™ EVOS™ M5000 Stage Plate
This EVOS M5000 Stage Plate accommodates a wide range of multi-well plates and all EVOS vessel holders designed for a mechanical stage.
Marca: Invitrogen™ AMEPVH089
Dettagli aggiuntivi : Peso : 0.68000kg
Descrizione
Individual vessel holders are readily switched, making it easy to image samples in a wide range of vessel formats. Vessel holders are available for microscope slides, cell culture flasks, Petri dishes, microwell plates, and more.
The EVOS M5000 Stage Plate offers these advantages:
• Perfect fit of multi-well plates
• Compatible adapters for a wide-range of vessel holders
• Plates and holders are easily changed
Specifica
EVOS M5000 | |
Stage Plate | |
Room Temperature |
EVOS | |
Cellular Imaging |
For Research Use Only. Not for use in diagnostic procedures.