Learn More
Invitrogen™ EVOS™ Onstage Two-port Perfusion Accessory
The EVOS™ perfusion accessory enables perfusion of a sample in the EVOS Onstage Incubator through two tube ports
Marca:  Invitrogen™ AMEP4769
Dettagli aggiuntivi : Peso : 0.09100kg
Codice prodotto. 17158865
  • 450.00€ / Pezzo
Spedizione stimata: 23-07-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
The EVOS Onstage Incubator is an environmental chamber designed specifically for the EVOS FL Auto Imaging System. When in use the perfusion accessory replaces the EVOS Onstage Incubator hose.
The EVOS Onstage Two-port Perfusion Accessory offers you these advantages:
• Ability to connect perfusion lines to the EVOS Onstage Incubator
• Facilitation of media exchanges
• Enhanced viability of sensitive cell types
• Extended live cell imaging duration
Designed for live cell imaging
The two ports on the perfusion accessory accommodate standard 1/8-in tubing, which is directed into the vessel on the stage. Perfusion of the sample with fresh medium or other liquids is generally done using a syringe or peristaltic pump (not included).
The EVOS Onstage Incubator is operated through the interface of the EVOS FL Auto Imaging system, and all environmental and acquisition parameters are controlled through this interface. So the incubator and imaging system operate as one fully integrated unit, making imaging live cells over time more convenient. Further, the heated lid of the EVOS Onstage Incubator helps prevent condensation and a glass coating of indium tin oxide maximizes light transmission.
Explore the entire EVOS line of imaging systems and accessories
Specifica
Specifica
Perfusion Accessory
EVOS
Room Temperature