Learn More
Thermo Scientific™ EZ-Link™ Pentylamine-Biotin
Biotinylate proteins or surfaces using EDC or other amine-reactive crosslinkers with this short (18.9A) biotinylation reagent.
Marca: Thermo Scientific™ 21345
Dettagli aggiuntivi : CAS : 67398-11-2 Peso : 0.09000kg
Descrizione
Thermo Scientific™ EZ-Link Pentylamine-Biotin is biotin that has been conjugated to pentylenediamine to add a short, amine-terminated spacer to its valeric acid group, providing a simple amine-modified compound for different applications.
Highlights:
- Requires the use of EDC crosslinking agent to couple to carboxylic acid groups or 5' phosphate groups
- Relatively water-soluble
- Forms an amide bond
Specifica
Pentylamine-Biotin | |
Store at 4°C. | |
Cell-Impermeant | |
Amine | |
Biotin | |
Biotin & Analogs |
50 mg | |
EZ-Link™ | |
Carboxylic Acid, Ketone, Aldehyde | |
Water | |
Short |
For Research Use Only. Not for use in diagnostic procedures.