Learn More
3M™ Face Shield 5E-11
326.00 EUR valido fino al 2024-07-31
Usa il codice promo "21674" per avere il tuo prezzo promozionale.
Usa il codice promo "21674" per avere il tuo prezzo promozionale.
Click to view available options
Frame Color: Green
Descrizione
- Shade 5 visor is ideal for outdoor use in bright light conditions
- Faceshield protects against UV, IR or visible light
- Compatible with the 3M™ V5 Face Shield system for mounting on a 3M™ Hard Hats Series G3000 or G22
- Snap in features makes it quick and easy to combine the face shield to the face shield holder
- Curved shape for close fit to the face
- Anti-Fog and Anti-Scratch resistant coating
- Optical class 1 visor, low energy impact, B
Specifica
Certificazione/Conformità | CE Approved , EN 166: 2001 |
Forma | Curved |
Lens Tint | Shade 5 |
Product Type | Protective Visor |
Colore telaio | Green |
Materiale | Polycarbonate |