Learn More
Thermo Scientific™ CpG Methyltransferase (M.SssI)
Sfoglia Tutto Thermo Scientific ProdottiDescrizione
The enzyme methylates the C5 position on the base moiety of all cytosine nucleotides contained in unmethylated or hemimethylated double stranded DNA in a 5'-CpG-3' context.
M.SssI is specifically formulated for fast reaction times without compromising the reaction efficiency. The enzyme completes modification of all CpGs in 15 minutes at 37°C. In addition, the enzyme has been specifically validated for use with genomic DNA -the primary substrate in epigenetic studies.
M.SssI is supplied with an optimized 10X reaction buffer and 50X S-adenosylmethionine (SAM) as a cofactor.
Highlights
- High efficiency -complete in vitro methylation of all CpGs in non-methylated and hemi-methylated DNA
- Fast -reaction is completed in 15 minutes at 37°C
- Stable -at -20°C for 2 years
Specifica
Specifica
| Enzyme | Methyltransferase |
| Compatible Buffer | 10X Reaction Buffer |
| Quantità | 25 μL |
| Product Type | CpG Methyltransferase |
For Research Use Only. Not for use in diagnostic procedures.
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.