Learn More
Coverguard™ FFP2 NR D Face Mask for Organic Vapors Gas and Acids
FFP2 half masks have an electrostatic filter media to filter bad odors. The design gives limited breathing resistance, thanks to the filtering material and exhalation valve with active carbon, aids breathing and avoids fog build-up inside the mask.
Marca: Coverguard™ 23236
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- PPE Cat. III FFP2 NR D Organic vapors, Gas and Acids
- Filters bad odors
- Exhalation valve with active carbon reduces fog build-up inside the mask
- Non-woven polypropylene
- Adjustable aluminum nose clip with PU foam total nose seal
- Specific design gives limited breathing resistance thanks to the large surface of the filtering material
Specifica
CE PPE Cat. III, FFP2 NR D, EN 149:2001 + A1:2009 | |
Single-use | |
Heavy & Process Industries, Chemical and Pharmaceutical Industries, Light Industries, Infrastructure, Building Industry, Construction - Finishing Work, DIY | |
Organic Vapors, Gas and Acids | |
Non-sterile |
FFP2 NR D Half Mask with Valve for Organic Vapors, Gas and Acids | |
Yes | |
Aluminum Nose Clip, Total PU Nose Seal | |
Half Mask | |
Half Mask with Shell and Carbon Valve |