Learn More
Lenz Laborglasintrumente Filter Funnels
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca: Lenz Laborglasintrumente 07195170
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Filter funnels
- Material: borosilicate glass 3.3
Specifica
1 L | |
320 mm | |
120 mm |
Borosilicate Glass (3.3) | |
120 mm | |
1 |