Learn More
Lenz Laborglasintrumente Filter Funnels
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca: Lenz Laborglasintrumente 07195228
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Filter funnels
- Material: borosilicate glass 3.3
Specifica
50 mL | |
30 mm | |
2 |
Borosilicate Glass (3.3) | |
35 mm |