Learn More
Hamilton™ Filter tube, Autolys, for automated lysis with 2D barcode on the tube bottom, with cap for automated processing, sterile
Marca:  Hamilton™ 235478
Dettagli aggiuntivi : Peso : 0.50000kg
Codice prodotto. 15177376
  • 1675.00€ / pezzi 500
Spedizione stimata: 10-09-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
The Hamilton consumables range is produced in an ISO Class 8 Clean Room, based on ISO 14644 standards as well as ISO 9001 and ISO 13485. Hamilton goes beyond relying on just statistical analysis of quality to assure the highest product quality, e.g. each CO-RE tip is checked for concentricity of its orifice and the absence of flash from the moulding process. TüV Rheinland certifies that our installed Quality Management System has been assessed and found to be in accordance with the requirements of the high performance standards for design and manufacturing of disposable products. Most consumables meet the EC directives CE and IVD. To meet the highly sophisticated needs of laboratories in the Genomics, Proteomics, Cellomics, Forensic and Drug Discovery fields, Hamilton has considerably refined the production requirements for sterile/non-sterile disposable products. The superior biological purity of CO-RE pipettor tips, tubes, and reagent containers is guaranteed by a controlled production process. Skilled personnel, along with automated clean room production, enable contamination-free manufacturing. At regular intervals the production batch is subjected to a highly sensitive PCR method to secure compliance with the defined purity criteria of Dnase/Rnase, pyrogen and PCR inhibitor-free. Hamilton has introduced a new purity grade especially for forensic work, to eliminate the residual risk of contamination of clean products. Along with the strictest purity requirements during the production process, an additional step is performed during the manufacturing and packing of disposable end products. This treatment ensures the effective removal of contaminants by physically destroying any DNA molecules. In accordance with Forensic Science International (FSI), Hamilton has validated the treatment process to prove the effectiveness of eliminating amplifiable DNA. Considering that just one contamination event from extraneous DNA during forensic investigation may result in incorrect DNA profiling, rendering evidence worthless, Hamilton's commitment to purity is of the utmost importance.
AutoLys tubes carry out the preparation of cleared lysates from various biological samples such as swabs, cloth cuttings, paper punches, human and animal tissues, plant material, etc. prior to purification of DNA, RNA and other biological molecules. Hamilton offers the unique AutoLys tube as an important accessory for the AutoLys STAR or as a standalone consumable, to minimise time-consuming lysis steps, provide full ID traceability, optimise yields, and increase reproducibility and productivity.
  • Revolutionary automated lysis system
  • Secure sample lysis and separation in one tube
  • Carry-over prevention
  • Full sample ID traceability
  • Minimises sample evaporation
  • Time saving
  • Hamilton purity concept certified and biological purity PLUS tested products including human DNA-free, Dnase-/Rnase-free, Pyrogen-free (endotoxin-free) and PCR inhibitor-free materials
  • Automation (A) and manual (M) type tubes available