Learn More
Thermo Scientific™ Pipette multicanale Finnpipette ™F1
Click to view available options
Volume (sistema metrico):
μ
5 to 50 μL
Da 10 a 100 μl
Da 30 a 300 μl
Numero di canali:
8
12
16
Colore:
Arancione
Giallo
Purple
Rosa
Turchese
Dimensione della confezione:
Pezzo
Descrizione
I livelli di prestazioni e comfort ergonomico fanno delle pipette multicanale Thermo Scientific™ Finnpipette™ F1 la scelta ideale per ogni laboratorio. Disponibili modelli a 8, 12 o 16 canali, è possibile scegliere tra una vasta gamma di puntali per pipette Finntip per risultati ottimali.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F1: Everything you want in your everyday, go-to pipette
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI
Silent, Effortless Volume Adjustment
Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.
Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50 μl models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
CE marked
The Finnpipette F1 Pipette together with compatible Finntip tips is CE marked in accordance with IVD Directive in Europe.
5 Year Warranty
With convenient online registration, and may vary by country.
Learn more about the complete Finnpipette family >
Finnpipette F1: Everything you want in your everyday, go-to pipette
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI
Silent, Effortless Volume Adjustment
Enjoy the new silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models.
Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50 μl models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
CE marked
The Finnpipette F1 Pipette together with compatible Finntip tips is CE marked in accordance with IVD Directive in Europe.
5 Year Warranty
With convenient online registration, and may vary by country.
Learn more about the complete Finnpipette family >
Specifica
Specifica
| Punte compatibili | Finntip 20 Micro, Filter 20 Micro |
| Descrizione | Finnpipette F1 |
| Colore | Purple |
| Numero di canali | 16 |
| Volume (sistema metrico) | μ |
| Incrementi | 0.02μL |
Conformi alla direttiva IVD 98/79/CE, valida solo in Europa. Non idonee per la diagnostica in vitro in Nord America.