Learn More
Thermo Scientific™ Finnpipette™ F2 Variable Volume Pipettes
GREENER_CHOICE
Codice prodotto. p-4530188
Click to view available options
Volume (sistema metrico):
μ
1 to 10 mL
5 to 50 μL
Da 0.2 a 2 µl, micro
Da 0.5 a 5 ml
Da 0.5 a 5 µl, micro
Da 10 a 100 μl
Da 2 a 20 µl
Da 2 a 20 µl, micro
Da 20 a 200 μl
Da 5 a 50 µl
Punte compatibili:
10 ml, Flex 10 Est.
5 ml
Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
Finntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000
Finntip 250, Filter 10, Flex 200, Flex Filter 30, 200 Ext, Wide 250
Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, Wide 250
Finntip 250, Filter 20, Flex 200, Flex Filter 30, 200 Ext, Wide 250
Finntip 250, Filter 200, Flex 200, Flex Filter 200, 200 Ext, Filter 200 Ext, Wide 250
Finntip 50 Micro, Filter 50 Micro
Finntip Flex 200, Finntip Flex 300, Finntip 250 universale, Finntip 200 Extended
Flex 10, 10, 50
Colore:
Blu
Giallo
Rosa
Rosso
Turchese
Verde
Incrementi:
0.002 µl
0.01 µl
0.02 µl
0.02µL
μ
0,2 μl
1 µl
0.01 ml
0.02 ml
Dimensione della confezione:
Pezzo
Codice prodotto. 11877341
Marca: Thermo Scientific™ 4642010
Solo null disponibili
Aggiungi al carrello
Edge
Aggiungi al carrello
Meet the challenges of highly demanding applications where robustness and durability are essential with the sleek, rugged design of these fully autoclavable pipettes.
Meet the challenges of highly demanding applications where robustness and durability are essential with the sleek, rugged design of Thermo Scientific™ Finnpipette™ F2 Variable Volume Pipettes. Built with tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light, these variable volume pipettes are fully autoclavable without disassembling. They feature a wide selection of expressly designed tips for optimal performance, precision and accuracy.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F2: Proven reliable for long-term intensive use
Rugged and Durable
Finnpipette F2 contains tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Its rugged design withstands physical use without damage.
Fully autoclavable
Decontamination of the Finnpipette F2 is simple as there is no need to disassemble the F2 for autoclaving, minimizing disruption and downtime.
Maintenance is Easy
Simply detach the tip cone for efficient daily maintenance or decontamination when not using an autoclave.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50-μL models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
CE marked
The Finnpipette F2 Pipette together with compatible Finntip tips is CE marked in accordance with IVD Directive in Europe.
5 Year Warranty
With convenient online registration, and may vary by country.
TRUSTED_SUSTAINABILITY
Specifica
Punte compatibili Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10
Descrizione Finnpipette F2
Autoclavabile Autoclavable
Colore Rosa
Numero di canali 1
Product Line Finnpipette™ F2
Tipo Variable Volume Pipette
Volume (sistema metrico) Da 0.2 a 2 µl, micro
Incrementi 0.002 µl
Codice colore Pink